missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCBP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10349-1-AP-150UL
This item is not returnable.
View return policy
Description
The product of this gene is a component of the nuclear cap-binding protein complex, which binds to the monomethylated 5′ cap of nascent pre-mRNA in the nucleoplasm. The encoded protein promotes high-affinity mRNA-cap binding and associates with the CTD of RNA polymerase II. The CBC promotes pre-mRNA splicing, 3′-end processing, RNA nuclear export, and nonsense-mediated mRNA decay.Specifications
| NCBP1 | |
| Polyclonal | |
| Unconjugated | |
| NCBP1 | |
| CBP80, NCBP, NCBP 80 kDa subunit, NCBP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 433702, 4686 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q09161, Q3UYV9 | |
| Ncbp1 | |
| NCBP1 Fusion Protein Ag0496 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction