missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCBP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13454637
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13454637 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13454637 Supplier Invitrogen™ Supplier No. PA530098

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (81%), Chicken (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May be involved in protein trafficking from the plasma membrane to the early endosome (EE) as well as in homotypic fusion of endocytic organelles. Mediates the endocytic trafficking from early endosomes to late endosomes and lysosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCBP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene NCBP1
Gene Accession No. Q09161, Q3UYV9
Gene Alias 80 kDa nuclear cap-binding protein; AU014645; AW538051; CBP80; MGC2087; NCBP; NCBP 80 kDa subunit; Ncbp1; nuclear cap binding protein subunit 1; nuclear cap binding protein subunit 1, 80kD; nuclear cap binding protein subunit 1, 80kDa; nuclear cap-binding protein subunit 1; RP11-546O6.1; Sto1
Gene Symbols NCBP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human CBP80. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 433702, 4686
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.