missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528417
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Chicken (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in protein trafficking from the plasma membrane to the early endosome (EE) as well as in homotypic fusion of endocytic organelles. Mediates the endocytic trafficking from early endosomes to late endosomes and lysosomes.
Specifications
| NCBP1 | |
| Polyclonal | |
| Unconjugated | |
| NCBP1 | |
| 80 kDa nuclear cap-binding protein; AU014645; AW538051; CBP80; MGC2087; NCBP; NCBP 80 kDa subunit; Ncbp1; nuclear cap binding protein subunit 1; nuclear cap binding protein subunit 1, 80kD; nuclear cap binding protein subunit 1, 80kDa; nuclear cap-binding protein subunit 1; RP11-546O6.1; Sto1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4686 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q09161 | |
| NCBP1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human CBP80. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction