missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCBP1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41222CL1
This item is not returnable.
View return policy
Description
NCBP1 Polyclonal antibody specifically detects NCBP1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| NCBP1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 4686 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| CBP80nuclear cap binding protein subunit 1, 80kD, NCBP 80 kDa subunit, NCBPMGC2087, nuclear cap binding protein subunit 1, 80kDa, nuclear cap-binding protein subunit 1,80 kDa nuclear cap-binding protein, Sto1 | |
| Antibody was raised against a 17 amino acid peptide near the center of human NCBP1. The immunogen is located within amino acids 420 - 470 of NCBP1. Amino Acid Squence: LSQDPESPKPKFVREV | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction