missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCAPH Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA551727

Product Code. 15769680

  • £483.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LHCQDYRSEL LFPSDVQTLS TGEPLELPEL GCVEMTDLKA PLQQCAEDRQ ICPSLAGFQF TQWDSETHNE SVSALVDKFK KNDQVFDINA EVDESDCGDF PDGSLGDDFD ANDEPDHT Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 82%.

This gene encodes a member of the barr gene family and a regulatory subunit of the condensin complex. This complex is required for the conversion of interphase chromatin into condensed chromosomes. The protein encoded by this gene is associated with mitotic chromosomes, except during the early phase of chromosome condensation. During interphase, the protein has a distinct punctate nucleolar localization.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NCAPH
Polyclonal
Unconjugated
NCAPH
A730011O11Rik; barren homolog 1; barren homolog protein 1; Brrn; BRRN1; Caph; CAP-H; Chromosome-associated protein H; condensin complex subunit 2; HCAP-H; KIAA0074; mCAP-H; mKIAA0074; Ncaph; Non-SMC condensin I complex subunit H; non-SMC condensin I complex, subunit H; XCAP-H homolog
Rabbit
Antigen affinity chromatography
RUO
23397
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q15003
NCAPH
Recombinant protein corresponding to Human NCAPH. Recombinant protein control fragment (Product #RP-102187).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.