missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ NCAPG2 Recombinant Protein Antigen

Product Code. 18345033 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345033 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345033 Supplier Novus Biologicals™ Supplier No. NBP317400PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 54892
Molecular Weight (g/mol) M.W. Theoretical: 29 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias Chromosome-associated protein G2, FLJ20311, Leucine zipper protein 5CAP-G2hCAP-G2condensin-2 complex subunit G2, LUZP5, more than blood homolog, MTB, Non-SMC condensin II complex subunit G2, non-SMC condensin II complex, subunit G2
Gene Symbol NCAPG2
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein NCAPG2
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.