missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCAPG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101540
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NCAPG.
This gene encodes a subunit of the condensin complex, which is responsible for the condensation and stabilization of chromosomes during mitosis and meiosis. Phosphorylation of the encoded protein activates the condensin complex. There are pseudogenes for this gene on chromosomes 8 and 15. Alternative splicing results in multiple transcript variants.
Specifications
| NCAPG | |
| Polyclonal | |
| Unconjugated | |
| NCAPG | |
| 5730507H05Rik; CAPG; CHCG; chromosome condensation protein G; chromosome-associated protein G; Condensin complex subunit 3; condensin subunit CAP-G; FLJ12450; Hcapg; hCAP-G; Melanoma antigen NY-MEL-3; MFT.M05.13; MGC126525; NCAPG; Non-SMC condensin I complex subunit G; non-SMC condensin I complex, subunit G; NYMEL3; NY-MEL-3; on-SMC condensin I complex, subunit G; RGD1562646; XCAP-G homolog; YCG1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54392, 64151 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BPX3 | |
| NCAPG | |
| A synthesized peptide derived from human NCAPG(Accession Q9BPX3), corresponding to amino acid residues N950-E1000. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction