missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCAPD3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15694295
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15694295 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15694295 Supplier Invitrogen™ Supplier No. PA550368

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total NCAPD3 protein.

This protein is a regulatory subunit of the condensin-2 complex, a complex which establishes mitotic chromosome architecture and is involved in physical rigidity of the chromatid axis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAPD3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene NCAPD3
Gene Accession No. P42695
Gene Alias 2810487N22Rik; 4632407J06Rik; AI195468; AU018739; B130055D15Rik; CAPD3; CAP-D3; Condensin-2 complex subunit D3; hCAP-D3; hcp-6; hHCP-6; Jacobsen syndrome region 2; Kiaa0056; mKIAA0056; NCAPD3; Non-SMC condensin II complex subunit D3; non-SMC condensin II complex, subunit D3
Gene Symbols NCAPD3
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the C terminal of human non-SMC condensin II complex, subunit D3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23310
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.