missing translation for 'onlineSavingsMsg'
Learn More

NCAPD3 Antibody (1D5), Novus Biologicals™

Product Code. 18386998 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386998 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386998 Supplier Novus Biologicals Supplier No. H00023310M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NCAPD3 Monoclonal antibody specifically detects NCAPD3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAPD3
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1D5
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH11408
Gene Alias CAPD3, CAP-D3, FLJ42888, hCAP-D3condensin-2 complex subunit D3, hcp-6, hHCP-6, KIAA0056MGC104671, Non-SMC condensin II complex subunit D3, non-SMC condensin II complex, subunit D3
Host Species Mouse
Immunogen KIAA0056 (AAH11408.1, 1 a.a. ~ 341 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRSKPDKDLLMEEDDMALANVVMQEAQKKLISQVQKRNFIENIIPIIISLKTVLEKNKIPALRELMHYLREVMQDYRDELKDFFAVDKQLASELEYDMKKYQEQLVQEQELAKHADVAGTAGGAEVAPVAQVALCLETVPVPAGQENPAMSPAVSQPCTPRASAGHVAVSSPTPETGPLQRLLPKARPMSLSTIAILNSVKKAVESKSRHRSRSLGVLPFTLNSGSPEKTCSQVSSYSLEQESNGEIEHVTKRAI
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 23310
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.