missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCAPD2 Recombinant Rabbit Monoclonal Antibody (24GB75)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£172.00 - £488.00

Specifications

Antigen NCAPD2
Clone 24GB75
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276468
GREENER_CHOICE
View Documents
Invitrogen™
MA557514
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275282
View Documents
Invitrogen™
MA557515
20 μL
£172.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NCAPD2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Regulatory subunit of the condensin complex, a complex required for conversion of interphase chromatin into mitotic-like condense chromosomes. The condensin complex probably introduces positive supercoils into relaxed DNA in the presence of type I topoisomerases and converts nicked DNA into positive knotted forms in the presence of type II topoisomerases. May target the condensin complex to DNA via its C-terminal domain (PubMed:11136719). May promote the resolution of double-strand DNA catenanes (intertwines) between sister chromatids. Condensin-mediated compaction likely increases tension in catenated sister chromatids, providing directionality for type II topoisomerase-mediated strand exchanges toward chromatid decatenation. Required for decatenation of non-centromeric ultrafine DNA bridges during anaphase. Early in neurogenesis, may play an essential role to ensure accurate mitotic chromosome condensation in neuron stem cells, ultimately affecting neuron pool and cortex size (PubMed:27737959). [UniProt]
Specifications

Specifications

NCAPD2
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
NCAPD2
2810406C15Rik; 2810465G24Rik; Capd2; CAP-D2; chromosome condensation related SMC associated protein 1; chromosome condensation-related SMC-associated protein 1; Chromosome-associated protein D2; CNAP1; Condensin complex subunit 1; hCAP-D2; KIAA0159; mCAP-D2; mKIAA0159; Ncapd2; Non-SMC condensin I complex subunit D2; non-SMC condensin I complex, subunit D2; RGD1562596; XCAP-D2 homolog
NCAPD2
Primary
Affinity chromatography
24GB75
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q15021, Q8K2Z4
362438, 68298, 9918
A synthesized peptide derived from human CNAP1 (aa 100-200).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.