missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCAM2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5110015

Product Code. 17177943

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: YGVKIIELSQ TTAKVSFNKP DSHGGVPIHH YQVDVKEVAS EIWKIVRSHG VQTMVVLNNL EPNTTYEIRV AAVNGKGQGD YSKIEIFQTL PVREPSPPSI HGQPSSGKSF KLSITKQDDG GAPILEYIVK YRSKDKEDQW LEKKVQGNKD HIILEHLQWT MGYEVQITAA NRLGYSEPTV YEFSMPPKPN I.

NCAM2 (neural cell adhesion molecule 2) is a cell adhesion molecule belonging to the immunoglobulin superfamily. It is differentially expressed in the developing and mature olfactory epithelium and is involved in the guidance of developing olfactory sensory neurons to the glomeruli of the olfactory bulb.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NCAM2
Polyclonal
Unconjugated
NCAM2
fasciclin II; NCAM2; N-CAM-2; Ncam-2; NCAM21; neural cell adhesion molecule 2; Neural cell adhesion molecule RB-8; Ocam; OCAM-GPI; olfactory axon cell adhesion molecule; R4B12; R4B12 antigen; Rncam
Rabbit
Affinity chromatography
RUO
17968, 288280, 4685
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.64 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15394, O35136
NCAM2
Recombinant fusion protein containing a sequence corresponding to amino acids 500-690 of human NCAM2 (NP_004531.2).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.