missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCAM2 Monoclonal Antibody (125101)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561866
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
NCAM2 (neural cell adhesion molecule 2) is a cell adhesion molecule belonging to the immunoglobulin superfamily. It is differentially expressed in the developing and mature olfactory epithelium and is involved in the guidance of developing olfactory sensory neurons to the glomeruli of the olfactory bulb.
Specifications
| NCAM2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O35136 | |
| NCAM2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse OCAM/NCAM2, Leu20-Gly700 (predicted). | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 125101 | |
| Unconjugated | |
| NCAM2 | |
| fasciclin II; NCAM2; N-CAM-2; Ncam-2; NCAM21; neural cell adhesion molecule 2; Neural cell adhesion molecule RB-8; Ocam; OCAM-GPI; olfactory axon cell adhesion molecule; R4B12; R4B12 antigen; Rncam | |
| Rat | |
| Protein A/G | |
| RUO | |
| 17968 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction