missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAM1 Mouse anti-Human, Clone: NCAM1/795, Abnova™
Mouse monoclonal antibody raised against full length recombinant human NCAM1.
Brand: Abnova MAB13381.100ug
This item is not returnable.
View return policy
Specifications
| NCAM1 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determ | |
| CD56/MSK39/NCAM | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 4684 | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| NCAM1/795 | |
| Mouse monoclonal antibody raised against full length recombinant human NCAM1. | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
| NCAM1 | |
| Recombinant protein corresponding to full length human NCAM1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction