missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAM-1/CD56 Antibody (735), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52710PECY55
This item is not returnable.
View return policy
Description
NCAM-1/CD56 Monoclonal specifically detects NCAM-1/CD56 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| NCAM-1/CD56 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| NCAM1 | |
| Homopolymers of alpha 2,8-linked sialic acid of Escherichia coli K1. | |
| 0.1 mL | |
| Primary | |
| Recognises Polysialic acid (PSA), a carbohydrate polymer attached to the neural cell adhesion molecule (NCAM), which shares molecular epitopes with homopolymers of (alpha)2,8-linked sialic acid of Escherichia coli K1. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 735 | |
| Flow Cytometry | |
| CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
| Mouse | |
| Protein A or G purified | |
| Astrocyte Markers, Cellular Markers, Cytokine Research, Cytoskeleton Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
| 4684.0 | |
| Human, Mouse, Rat | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction