missing translation for 'onlineSavingsMsg'
Learn More

NCAM-1/CD56 Antibody (2524F) [DyLight 755], Novus Biologicals™

Product Code. 30732873 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30732873 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732873 Supplier Novus Biologicals Supplier No. FAB24085Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NCAM-1/CD56 Monoclonal antibody specifically detects NCAM-1/CD56 in Human,Rat samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAM-1/CD56
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2524F
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias CD56, CD56/NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant human NCAM-1/CD56, Leu20-Pro603 and Glu636-Asn741, Accession # NP_001070150
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4684
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.