missing translation for 'onlineSavingsMsg'
Learn More

NCAM-1/CD56 Antibody (123A8 (same as 56C04)) - Azide and BSA Free, Novus Biologicals™

Product Code. 18411622 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411622 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411622 Supplier Novus Biologicals Supplier No. NBP2345180.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NCAM-1/CD56 Monoclonal antibody specifically detects NCAM-1/CD56 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Imaging Mass Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAM-1/CD56
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Imaging Mass Cytometry
Classification Monoclonal
Clone 123A8 (same as 56C04)
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin 0.5 to 1 μg/mL, Imaging Mass Cytometry
Formulation 10 mM PBS
Gene Alias CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM
Host Species Mouse
Immunogen Membrane preparation of a small cell lung carcinoma
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4684
Target Species Human
Content And Storage Store at -20°C to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.