missing translation for 'onlineSavingsMsg'
Learn More

NCALD Mouse anti-Human, Clone: 6B4E10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66088-1-IG-20UL

 View more versions of this product

Product Code. 16877645

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is specific to NCALD.

This gene encodes a member of the neuronal calcium sensor (NCS) family of calcium-binding proteins. The protein contains an N-terminal myristoylation signal and four EF-hand calcium binding loops. The protein is cytosolic at resting calcium levels; however, elevated intracellular calcium levels induce a conformational change that exposes the myristoyl group, resulting in protein association with membranes and partial co-localization with the perinuclear trans-golgi network. The protein is thought to be a regulator of G protein-coupled receptor signal transduction. Several alternatively spliced variants of this gene have been determined, all of which encode the same protein; additional variants may exist but their biological validity has not been determined.
Specifications

Specifications

NCALD
Monoclonal
1.05 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P61601
NCALD
NCALD Fusion Protein Ag18760
20 μL
Primary
Human
Antibody
IgG2a
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
6B4E10
Unconjugated
NCALD
NCALD, neurocalcin delta
Mouse
Protein A
RUO
83988
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.