missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCALD Mouse anti-Human, Clone: 6B4E10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66088-1-IG-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to NCALD.
This gene encodes a member of the neuronal calcium sensor (NCS) family of calcium-binding proteins. The protein contains an N-terminal myristoylation signal and four EF-hand calcium binding loops. The protein is cytosolic at resting calcium levels; however, elevated intracellular calcium levels induce a conformational change that exposes the myristoyl group, resulting in protein association with membranes and partial co-localization with the perinuclear trans-golgi network. The protein is thought to be a regulator of G protein-coupled receptor signal transduction. Several alternatively spliced variants of this gene have been determined, all of which encode the same protein; additional variants may exist but their biological validity has not been determined.Specifications
| NCALD | |
| Monoclonal | |
| 1.05 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61601 | |
| NCALD | |
| NCALD Fusion Protein Ag18760 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 6B4E10 | |
| Unconjugated | |
| NCALD | |
| NCALD, neurocalcin delta | |
| Mouse | |
| Protein A | |
| RUO | |
| 83988 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction