missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nbs1, p Ser343 Antibody (SR2302), Novus Biologicals™
Rabbit Monoclonal Antibody
£282.00 - £367.00
Specifications
| Antigen | Nbs1 |
|---|---|
| Clone | SR2302 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649144
|
Novus Biologicals
NBP3-22663-100ul |
100 μg |
£367.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602805
|
Novus Biologicals
NBP3-22663-50ul |
50 μg |
£282.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nbs1 Monoclonal antibody specifically detects Nbs1 in Human samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| Nbs1 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex | |
| A synthesized peptide derived from human phospho-Nbs1 [Ser343] (Uniprot #: O60934) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2302 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4683 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title