missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nbs1 Antibody (7E4A2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-26297
This item is not returnable.
View return policy
Description
Nbs1 Monoclonal specifically detects Nbs1 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| Nbs1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500 - 1:2000, Simple Western 1:50, Flow Cytometry 1:200 - 1:400, ELISA, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1-50 - 1:100, Immunohistochemistry-Paraffin, Knockout Validated | |
| ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex | |
| Mouse | |
| 85 kDa | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors | |
| 4683 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 α |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
| 7E4A2 | |
| Unconjugated | |
| O60934 | |
| NBN | |
| Nbs1 Antibody (7E4A2) was made to partial recombinant human NBS1 (between residues 400-650) expressed in E. coli [Uniprot O60934] | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Precipitation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction