missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NBR1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
£185.00 - £433.00
Specifications
| Antigen | NBR1 |
|---|---|
| Concentration | 1.24 mg/mL |
| Dilution | Western Blot, Simple Western 1:400, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100, Sandwich ELISA Capture reported by customer review |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18240965
|
Novus Biologicals
NBP1-71703SS |
0.025 mL |
£185.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18717993
|
Novus Biologicals
NBP1-71703 |
0.1 mL |
£433.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NBR1 Polyclonal specifically detects NBR1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| NBR1 | |
| Western Blot, Simple Western 1:400, ELISA, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100, Sandwich ELISA Capture reported by customer review | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| Q14596, P97432 | |
| 4077 | |
| A genomic peptide made to an internal region of the human NBR1 protein (within residues 50-250). [Swiss-Prot Q14596] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.24 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| Cell migration-inducing gene 19 protein, FLJ98272, KIAA0049CA125, M17S2FLJ55359,1A1-3B, Membrane component chromosome 17 surface marker 2,1A13B, membrane component, chromosome 17, surface marker 2 (ovarian carcinoma antigenCA125), migration-inducing protein 19, neighbor of BRCA1 gene 1, Neighbor of BRCA1 gene 1 protein, next to BRCA1 gene 1 protein, Protein 1A1-3B | |
| NBR1 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Immunogen has 75% identity to the rat protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title