missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NBR1 Monoclonal antibody specifically detects NBR1 in Human, Mouse samples. It is validated for Western Blot, Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunomicroscopy
Specifications
Specifications
| Antigen | NBR1 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunomicroscopy |
| Classification | Monoclonal |
| Clone | 6B11 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_005890 |
| Gene Alias | Cell migration-inducing gene 19 protein, FLJ98272, KIAA0049CA125, M17S2FLJ55359,1A1-3B, Membrane component chromosome 17 surface marker 2,1A13B, membrane component, chromosome 17, surface marker 2 (ovarian carcinoma antigenCA125), migration-inducing protein 19, neighbor of BRCA1 gene 1, Neighbor of BRCA1 gene 1 protein, next to BRCA1 gene 1 protein, Protein 1A1-3B |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?