missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NBQX
GREENER_CHOICE

Product Code. 30000718
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30000718

Brand: Tocris Bioscience™ 0373/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent AMPA antagonist; more selective than CNQX (Cat. No. 0190)

NBQX is a selective and competitive AMPA and kainate receptor antagonist (IC50 = 0.15 μM and 4.8 μM, respectively). NBQX blocks the antidepressant effects of 8-Hydroxy-DPAT hydrobromide (Cat. No. 0529), decreases mTOR and BDNF levels. NBQX is neuroprotective, anticonvulsant, antinociceptive and active in vivo.

NBQX disodium salt, a more water-soluble form of NBQX, also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118876-58-7
Molecular Formula C12H8N4O6S

Specifications

Quantity 10 mg
Formula Weight 336.28
Purity >98%
Molecular Weight (g/mol) 336.278
Chemical Name or Material NBQX
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.