missing translation for 'onlineSavingsMsg'
Learn More

NBQX, Thermo Scientific™

Product Code. 17770629 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17770629 100 mg 100mg
17760629 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17770629 Supplier Thermo Scientific Acros Supplier No. 329261000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material NBQX
CAS 118876-58-7
Synonym 2,3-Dihydroxy-6-nitro-7-sulphamoylbenzo[f]quinoxaline-2,3-dione; 6-Nitro-7-sulphamoylbenzo[f]quinoxaline-2,3-dione
Quantity 100 mg
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C12 H8 N4 O6 S
Physical Form Powder
Color Almost-white to Yellow
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.