missing translation for 'onlineSavingsMsg'
Learn More

NBQX, MedChemExpress

Product Code. p-200097901
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16416308 10 mg 10mg
16412427 50 mg 50mg
16408897 5 mg 5mg
16406636 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16416308 Supplier Medchem Express Supplier No. HY15068

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress NBQX (FG9202) is a highly selective and competitive AMPA receptor antagonist. NBQX has neuroprotective and anticonvulsant activity.

NBQX (FG9202) is a highly selective and competitive AMPA receptor antagonist. NBQX has neuroprotective and anticonvulsant activity.

Specifikationer

Chemical Name or Material NBQX
CAS 118876-58-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C12H8N4O6S
Quantity 10 mg
Synonym FG9202
Solubility Information DMSO : ≥ 75 mg/mL (223.03 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(N1)C(NC2=C1C3=C(C([N+]([O-])=O)=C2)C(S(N)(=O)=O)=CC=C3)=O
Molecular Weight (g/mol) 336.28
Formula Weight 336.28
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.