missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NBPF12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116360
This item is not returnable.
View return policy
Description
NBPF12 Polyclonal Antibody for Western Blot
This gene is a member of the neuroblastoma breakpoint family (NBPF) which consists of dozens of recently duplicated genes primarily located in segmental duplications on human chromosome 1. This gene family has experienced its greatest expansion within the human lineage and has expanded, to a lesser extent, among primates in general. Members of this gene family are characterized by tandemly repeated copies of DUF1220 protein domains. Gene copy number variations in the human chromosomal region 1q21.1, where most DUF1220 domains are located, have been implicated in a number of developmental and neurogenetic diseases such as microcephaly, macrocephaly, autism, schizophrenia, cognitive disability, congenital heart disease, neuroblastoma, and congenital kidney and urinary tract anomalies. Altered expression of some gene family members is associated with several types of cancer. This gene family contains numerous pseudogenes.
Specifications
| NBPF12 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q5TAG4 | |
| NBPF12 | |
| KLH conjugated synthetic peptide between 148-181 amino acids from the Central region of human NBPF12. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| NBPF12 | |
| chomosome one amplified sequence 1 cyclophilin; chromosome 1 amplified sequence 1; COAS1; KIAA1245; NBPF12; Neuroblastoma breakpoint family member 12; neuroblastoma breakpoint family, member 12 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 149013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction