missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NBPF Pan Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5101712

Product Code. 16678184

  • £448.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Antibody detects endogenous levels of total NBPF1/9/10/12/14/15/16/20.

This gene is a member of the neuroblastoma breakpoint family (NBPF) which consists of dozens of recently duplicated genes primarily located in segmental duplications on human chromosome 1. This gene family has experienced its greatest expansion within the human lineage and has expanded, to a lesser extent, among primates in general. Members of this gene family are characterized by tandemly repeated copies of DUF1220 protein domains. Gene copy number variations in the human chromosomal region 1q21.1, where most DUF1220 domains are located, have been implicated in a number of developmental and neurogenetic diseases such as microcephaly, macrocephaly, autism, schizophrenia, mental retardation, congenital heart disease, neuroblastoma, and congenital kidney and urinary tract anomalies. Altered expression of some gene family members is associated with several types of cancer. This gene family contains numerous pseudogenes. [provided by RefSeq, Apr 2013].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NBPF Pan
Polyclonal
Unconjugated
NBPF9
AB13; AB14; AB23; AB6; AD2; AE01; AE5; AG1; AG3; chomosome one amplified sequence 1 cyclophilin; chromosome 1 amplified sequence 1; COAS1; DJ328E19.C1.1; KIAA1245; KIAA1693; NBFP10; NBFP12; NBFP14; NBFP15; NBFP16; NBFP20; NBFP9; NBG; NBPF; NBPF1; NBPF10; NBPF12; NBPF14; NBPF15; NBPF16; NBPF20; NBPF9; Neuroblastoma breakpoint family member 1; neuroblastoma breakpoint family member 10; neuroblastoma breakpoint family member 12; neuroblastoma breakpoint family member 14; neuroblastoma breakpoint family member 15; Neuroblastoma breakpoint family member 16; neuroblastoma breakpoint family member 20; Neuroblastoma breakpoint family member 9; neuroblastoma breakpoint family, member 1; neuroblastoma breakpoint family, member 10; neuroblastoma breakpoint family, member 12; neuroblastoma breakpoint family, member 14; neuroblastoma breakpoint family, member 15; neuroblastoma breakpoint family, member 16; neuroblastoma breakpoint family, member 20; neuroblastoma breakpoint family, member 9
Rabbit
Affinity chromatography
RUO
100132406, 100288142, 149013, 25832, 284565, 400818, 55672
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P0DPF2, P0DPF3, Q3BBV0, Q5TAG4, Q5TI25, Q6P3W6, Q8N660
NBPF1, NBPF10, NBPF12, NBPF14, NBPF15, NBPF20, NBPF9
A synthesized peptide derived from human NBPF1(Accession Q3BBV0), corresponding to amino acid residues L1075-E1125.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.