Learn More
Tocris Bioscience™ NBOH-2C-CN hydrochloride
High affinity, selective 5-HT2A agonist
Specifications
Purity >98%
View More Specs
Description
Description
High affinity, selective 5-HT2A agonist (Ki = 1.3 nM). Exhibits 100-fold selectivity for 5-HT2A over 5-HT2C. Elicits head twitch behavior in mice.
Specifications
Specifications
>98%