missing translation for 'onlineSavingsMsg'
Learn More

NBN Mouse anti-Human, Clone:7E4C2, Abnova™

Mouse monoclonal antibody raised against recombinant human NBN.

Brand:  Abnova MAB17791.100ug

Product Code. 16095066

  • £535.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Mutations in this gene are associated with Nijmegen breakage syndrome, an autosomal recessive chromosomal instability syndrome characterized by microcephaly, growth retardation, immunodeficiency, and cancer predisposition. The encoded protein is a member of the MRE11/RAD50 double-strand break repair complex which consists of 5 proteins. This gene product is thought to be involved in DNA double-strand break repair and DNA damage-induced checkpoint activation. [provided by RefSeq]
Specifications

Specifications

NBN
Monoclonal
Unconjugated
ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry (1:200-1:1000)
Immunohistochemistry (1:200-1:1000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
NBN
Mouse
100 μg
Human
Antibody
IgG2a
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
7E4C2
nibrin
In PBS (0.05% sodium azide)
AT-V1, AT-V2, ATV, FLJ10155, MGC87362, NBS, NBS1, P95
Recombinant protein corresponding to amino acids 467-615 of human NBN from E. coli.
4683
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.