missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NBI 27914 hydrochloride
Description
Selective, non-peptide corticotropin-releasing factor1 (CRF1) receptor antagonist (Ki = 1.7 nM); has no activity at CRF2 receptors. Blocks behavioral seizures in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 470.66 |
| Purity | >99% |
| Molecular Weight (g/mol) | 470.644 |
| Chemical Name or Material | NBI 27914 hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?