missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NB thymosin beta Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1073
This item is not returnable.
View return policy
Description
NB thymosin beta Polyclonal specifically detects NB thymosin beta in Human, Mouse, Rat samples. It is validated for Peptide ELISA.
Specifications
| NB thymosin beta | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| NB thymosin beta, TbNB, thymosin beta 15a, thymosin beta-15A, Thymosin beta-15B, thymosin, beta, identified in neuroblastoma cells, thymosin-like 8, Thymosin-like protein 8, TMSB15, TMSB15B, TMSL8, TMSNBTb15 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11013 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:2000 | |
| NP_068832. | |
| TMSB15A | |
| Peptide with sequence SDKPDLSEVEKFDR-C corresponding to N-Terminus according to NP_068832.1. | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction