missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NB-598 Maleate, MedChemExpress
Description
NB-598 Maleate is a potent and competitive inhibitor of squalene epoxidase (SE), and suppresses triglyceride biosynthesis through the farnesol pathway.
Specifications
Specifications
| Chemical Name or Material | NB-598 Maleate |
| CAS | 155294-62-5 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C31H35NO5S2 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 100 mg/mL (176.76 mM; Need ultrasonic) |
| SMILES | O=C(O)/C=C\C(O)=O.CC(C)(C)C#C/C=C/CN(CC)CC1=CC(OCC2=CC(C3=CSC=C3)=CS2)=CC=C1 |
| Molecular Weight (g/mol) | 565.74 |
| Formula Weight | 565.74 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?