missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Naxitamab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559927
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Naxitamab is a humanized monoclonal antibody targeting the GD2 antigen. GD2 antigen is expressed on the surface of tumors generated by neuroectoderm, including neuroblastoma, melanoma and osteosarcoma. By binding to the GD2 antigen on the tumor surface, naxitamab can trigger antibody-mediated cytotoxicity and activate the complement system in the immune system to achieve the effect of killing tumors.
Specifications
| Naxitamab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Hu3F8 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction