missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAV3 Polyclonal Antibody
Description
This antibody detects endogenous protein at a molecular weight of 256 kDa. Purity is >95% by SDS-PAGE.
Neuron navigator 3 (NAV3), also known as pore membrane and/or filamentinteracting-like protein 1 (POMFIL1), unc-53 homolog 3 (unc53H3) or STEERIN3, is a 2,385 amino acid protein that localizes to the nuclear outer membrane and plays a role in neuron regeneration. Existing as three alternatively spliced isoforms, Neuron navigator 3 is highly expressed in adult and fetal brain and is found at moderate to low levels in ovary, lung, testis, placenta and heart. Neuron navigator 3 is thought be involved in the regulation of IL-2 production and contains one CH (calponin-homology) domain, three N-terminal hydrophobic domains, multiple N-glycosylation sites, three ATP/GTP-binding A motifs (P loops) and several phosphorylation sites.
Specifications
Specifications
| Antigen | NAV3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | NAV3 |
| Gene Accession No. | Q80TN7, Q8IVL0 |
| Gene Alias | 4732483H20Rik; 9630020C08Rik; KIAA0938; mKIAA0938; NAV3; Neuron navigator 3; POMFIL1; Pomfil1p; pore membrane and/or filament interacting like protein 1; pore membrane and/or filament-interacting-like protein 1; RGD1306259; steerin 3; STEERIN3; Steerin-3; Unc-53 homolog 3; unc53H3 |
| Gene Symbols | NAV3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?