missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAV2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £470.00
Specifications
| Antigen | NAV2 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18457630
|
Novus Biologicals
NBP1-84616-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18228187
|
Novus Biologicals
NBP1-84616 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NAV2 Polyclonal specifically detects NAV2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NAV2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 89797 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LREPSKTALGSSLPGLVNQTDKEKGISSDNESVASCNSVKVNPAAQPVSSPAQTSLQPGAKYPDVASPTLRRLFGGKPTKQVPIATAENMKNSVVISNPHATMTQQGNLDSPSGSGVLSSGSSSPLYSKNVDLNQSPL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| EC 3.6.4.12, FLJ10633, FLJ11030, FLJ23707, HELAD1UNC53H2, Helicase APC down-regulated 1, KIAA1419STEERIN2, neuron navigator 2, POMFIL2helicase, APC down-regulated 1, pore membrane and/or filament interacting like protein 2, Pore membrane and/or filament-interacting-like protein 2, RAINB1FLJ77876, retinoic acid inducible gene in neuroblastoma 1, Retinoic acid inducible in neuroblastoma 1, Steerin-2, Unc-53 homolog 2, unc53H2 | |
| NAV2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title