Learn More
Novus Biologicals™ Nav1.7 Antibody (Duke anti-NAv1.7) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-27987-1mg
 View more versions of this product
Product Code. 30044922
  • £695.00 / 1mg
Description
Nav1.7 Monoclonal antibody specifically detects Nav1.7 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nav1.7
Monoclonal
Unconjugated
Q15858
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Duke anti-NAv1.7
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
ETHA, FEB3B, NE-NA, NENAVoltage-gated sodium channel subunit alpha Nav1.7, Neuroendocrine sodium channel, Peripheral sodium channel 1, PN1hNE-Na, sodium channel protein type 9 subunit alpha, Sodium channel protein type IX subunit alpha, sodium channel, voltage-gated, type IX, alpha subunit, voltage-gated sodium channel alpha subunit Nav1.7, voltage-gated, type IX, alpha polypeptide
SCN9a/ Nav1.7
1 mg
Cancer, Cellular Signaling, Neuroscience
6335
Human
Purified