missing translation for 'onlineSavingsMsg'
Learn More

Nav1.6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18379346 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379346 100 μL 100µL
18323117 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18379346 Supplier Novus Biologicals Supplier No. NBP303464100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Nav1.6 Polyclonal specifically detects Nav1.6 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nav1.6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias CerIII, hNa6/Scn8a voltage-gated sodium channel, MED, NaCh6, Nav1.6, PN4, sodium channel protein type 8 subunit alpha, Sodium channel protein type VIII subunit alpha, sodium channel, voltage gated, type VIII, alpha polypeptide, sodium channel, voltage gated, type VIII, alpha subunit, Voltage-gated sodium channel subunit alpha Nav1.6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1350-1440 of human Nav1.6 (NP_055006.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6334
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.