missing translation for 'onlineSavingsMsg'
Learn More

Nav1.5 Antibody, Novus Biologicals™

Product Code. 18394220 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394220 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394220 Supplier Novus Biologicals Supplier No. NBP286726

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Nav1.5 Polyclonal specifically detects Nav1.5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nav1.5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose
Gene Alias cardiac sodium channel alpha subunit, cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit, CDCD2VF1, HB1sodium channel, voltage-gated, type V, alpha (long QT syndrome 3), HB2, HBBD, HH1CMD1E, ICCD, IVF, LQT3SSS1CMPD2, Nav1.5, PFHB1, Sodium channel protein cardiac muscle subunit alpha, sodium channel protein type 5 subunit alpha, sodium channel protein type V alpha subunit, Sodium channel protein type V subunit alpha, sodium channel, voltage-gated, type V, alpha subunit, Voltage-gated sodium channel subunit alpha Nav1.5
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of Human Nav1.5
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 6331
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.