missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NAV 26
GREENER_CHOICE

Product Code. p-5545914
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18774858 10 mg 10mg
18759248 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18774858 Supplier Tocris Bioscience™ Supplier No. 4691/10
Add to basket
Add to basket
This item is not returnable. View return policy

Selective Nav1.7 channel blocker

NAV 26 is a selective Nav1.7 channel blocker (IC50 = 370 nM). Exhibits >85-fold selectivity for Nav1.7 over Nav1.5 and hERG channels. Reduces nociceptive behavior in rat models of pain.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
Quantity 10 mg
Product Type NAV 26
Molecular Weight (g/mol) 434.41
Inhibitors NAV 26

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.