Learn More
Abeba™ Nature 8045 Shoes
Product Code. 15682988
Click to view available options
Size:
34
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15682988
Brand: Abeba™ 8045/39
Only null left
Add to basket
Add to basket
Leather covered cork footbed is ESD dissipative.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Black
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Adjustable instep area
  • Buckle fastening
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color Black
Footbed Material Cork
Material Leather
Size 39
Sole Features Slip-resistant
Sole Material Rubber
Style 8045