missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Natural Rubber Safety Pipette Fillers

Product Code. 10079780
Change view
Click to view available options
Type:
Safety Pipette Filler
Model:
Flip
Standard
Universal
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Model unitSize
10079780 Safety Pipette Filler Universal Each
10527451 Safety Pipette Filler Flip Each
10707281 Safety Pipette Filler Standard Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10079780 Supplier Hirschmann™ Supplier No. 9910103
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Made of natural rubber

  • Universal use for volumetric and graduated pipettes

Specifications

Color Red
Material Natural Rubber
Model Universal
Type Safety Pipette Filler
For Use With (Equipment) volumetric and graduated pipettes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.