Learn More
Masterflex™ Natural Polypropylene Barbed NPT Male Pipe Adapter Elbows
Specifications
Specifications
| Material | Polypropylene |
| For Use With | Fits 3/8 in. Tubing I.D. |
| Packaging Quantity | 10 Pack |
| Port Size | 1-1/2 x 1-19/32in. |
| Thread Size | 1/4 in. MNPT |
| Product Type | Barbed NPT Male Pipe Adapter Elbows |