Learn More
Ark Plas Products™ Natural Kynar Premium Grade Barbed Elbow Connectors
An excellent option for connecting tubing in the laboratory, with a pressure rating of up to 125 P.S.I. without clamps.
Brand: Ark Plas Products™ MCX14-DF0
Code : A0
Additional Details : Weight : 0.00990kg
Description
- Molded from virgin, lot-traceable material only
- Manufactured to the most exacting standards
- Made with critical use applications in mind
- No parting line on barb
Specifications
Barbed Elbow Connector | |
10 Pack | |
Natural |
Kynar | |
1/8 x 1/8 in. Barb |