Learn More
Fisherbrand™ Natural HDPE Cylinder Bottles with White Polypropylene Caps
High Density Polyethylene, natural color, good chemical resistance
Brand:  Fisherbrand™ FB02912112A
Product Code. 16194617
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Natural HDPE Narrow-Mouth Cylinder Bottles:
  • Shoulder style allows to pour small quantities without spilling
  • Ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping
Polypropylene PE Foam/PTFE Lined Caps:
  • Offers good impact strength, durability, thermal resistance, and pliability
  • Autoclavable
  • Liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • Liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference