missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Natriuretic Peptide Receptor C Antibody (OTI4C3), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Natriuretic Peptide Receptor C Monoclonal antibody specifically detects Natriuretic Peptide Receptor C in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | Natriuretic Peptide Receptor C |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI4C3 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ANP-C, ANPR-C, ANPRCFLJ14054, Atrial natriuretic peptide clearance receptor, atrial natriuretic peptide receptor 3, Atrial natriuretic peptide receptor type C, C5orf23, chromosome 5 open reading frame 23, GUCY2B, natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptidereceptor C), NPRC, NPR-C |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human NPR3 (NP_000899) produced in HEK293T cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?