missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Natriuretic Peptide Receptor C Antibody (1060830) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB102331JF525
This item is not returnable.
View return policy
Description
Natriuretic Peptide Receptor C Monoclonal antibody specifically detects Natriuretic Peptide Receptor C in Human samples. It is validated for Flow Cytometry,Western Blot
Specifications
| Natriuretic Peptide Receptor C | |
| Monoclonal | |
| Janelia Fluor 525 | |
| ANP-C, ANPR-C, ANPRCFLJ14054, Atrial natriuretic peptide clearance receptor, atrial natriuretic peptide receptor 3, Atrial natriuretic peptide receptor type C, C5orf23, chromosome 5 open reading frame 23, GUCY2B, natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptidereceptor C), NPRC, NPR-C | |
| Human embryonic kidney cell, HEK293-derived human NPRC/NPR3, Met1-Glu481, Accession # P17342 | |
| RUO | |
| 4883 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Western Blot | |
| 1060830 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction