Learn More
Abcam Native Staphylococcus aureus Protein A (HRP)
Product Code. 32143602 Shop All Abcam Products
Change view
Click to view available options
1 mg
Product Code.
32143602 1 mg
1 options
Product Code. 32143602 Supplier Abcam Supplier No. ab74561mg
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Native Staphylococcus aureus Protein A (HRP) is a Staphylococcus aureus subsp. aureus NCTC 8325 Full Length protein, expressed in Native, with >95%, suitable for Dot, IM, WB, ELISA, IHC-P.
Native Staphylococcus aureus Protein A (HRP) is a Staphylococcus aureus subsp. aureus NCTC 8325 Full Length protein, expressed in Native, with >95%, suitable for Dot, IM, WB, ELISA, IHC-P.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1 mg