missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Native Lipoteichoic Acid Mouse anti-Bacteria, Clone: mAb 55, Novus Biologicals™
Mouse Monoclonal Antibody
£163.00 - £426.00
Specifications
| Antigen | Native Lipoteichoic Acid |
|---|---|
| Clone | mAb 55 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:250, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay 1.2 ug/ml in PBS |
| Applications | Immunofluorescence, Immunohistochemistry, Western Blot, Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18040824
|
Novus Biologicals
NBP1-60146-0.025MG |
0.025 mg |
£163.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18749283
|
Novus Biologicals
NBP1-60146-0.1mg |
0.1 mg |
£426.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Native Lipoteichoic Acid Monoclonal specifically detects Native Lipoteichoic Acid in Bacteria samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay.Specifications
| Native Lipoteichoic Acid | |
| 0.2 mg/mL | |
| Immunofluorescence, Immunohistochemistry, Western Blot, Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Other | |
| LTA | |
| IgG3 | |
| Sterile filtered | |
| The specificity is labled not applicable. |
| mAb 55 | |
| Western Blot 1:1000, Flow Cytometry 1:250, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay 1.2 ug/ml in PBS | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS. 1 ml >200 ug/ml) culture medium with a low endotoxin level with 0.02% Sodium Azide | |
| Microbial mixture of Streptococcus sobrims HG961, HG962, HG970, and HG977 | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title