missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Native Lipoteichoic Acid Antibody (55), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62811
This item is not returnable.
View return policy
Description
Native Lipoteichoic Acid Monoclonal specifically detects Native Lipoteichoic Acid in Bacterial samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay.
Specifications
| NativeLipoteichoicAcid | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.1% BSA with 0.02% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Store at 4°C. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), In Situ Hybridization (ISH), Western Blot | |
| 55 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay | |
| LTA | |
| Microbial mixture of Streptococcus sobrims HG961, HG962, HG970 and HG977 | |
| 100 μg | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction