missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ National™ Target™ Kim-Snap™ Closures
Shop All Thermo Scientific Products
Click to view available options
Color:
Black
Yellow
Material (Liner):
PTFE/Rubber
PTFE/Silicone
Unit Size:
Pack of 100
Description
- Effective evaporation resistance—apply with simple thumb pressure
- Replace aluminum crimp seals and end tedious crimping/de-crimping
Specifications
Specifications
| Material (Liner) | PTFE/Rubber |
| Color | Yellow |
| Material | Polypropylene |
| Closure Size | 11 mm |
| For Use With (Equipment) | Target Snap/Crimp LoVial Vials |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction